missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MMP7 Monoclonal Antibody (J9), Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

£161.00 - £333.00

Specifications

Antigen MMP7
Clone J9
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17958802
View Documents
Invitrogen™
MA551010
20 μL
£161.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17908692
GREENER_CHOICE
View Documents
Invitrogen™
MA551009
200 μL
£333.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The enzyme encoded by this gene degrades proteoglycans, fibronectin, elastin and casein and differs from most MMP family members in that it lacks a conserved C-terminal protein domain. The enzyme is involved in wound healing, and studies in mice suggest that it regulates the activity of defensins in intestinal mucosa. The gene is part of a cluster of MMP genes which localize to chromosome 11q22. 3.
Specifications

Specifications

MMP7
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
HGNC:7174; MAT; Matrilysin; matrilysin, uterine; matrin; matrix metallo protease; matrix metallopeptidase 7; matrix metallopeptidase 7 (matrilysin, uterine); matrix metalloproteinase 7; Matrix metalloproteinase 7 (matrilysin); matrix metalloproteinase 7 (matrilysin, uterine); matrix metalloproteinase-7; MMP; MMP 7; MMP7; Mmp-7; MMP-7 protein; MMPs; MPMM; MPSL1; PUMP 1; Pump 1 protease; PUMP1; PUMP-1; pump-1 protease; uterine matrilysin; uterine metalloproteinase
MMP7
IgG1 κ
Protein A/G
MMP7
J9
Immunohistochemistry (Paraffin), Western Blot, Western Blot
Unconjugated
Mouse
Human, Pig
P09237
397411, 4316
Recombinant Human MMP7 protein, Leu18-Lys267 (Accession #P09237).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.