missing translation for 'onlineSavingsMsg'
Learn More

MMP3 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1F5A9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66338-1-IG-20UL

 View more versions of this product

Product Code. 16818125

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MMP3 (stromelysin-1) belongs to the peptidase M10A subfamily. It is secreted in its pro-protein form, and is activated by extracellular proteinase cleavage. MMP3 is capable of binding to zinc and calcium ions, and has metallo-endopeptidase activity. The active enzyme degrades collagen, gelatin, fibronectin, and laminin. It is involved in normal cellular processes such as wound repair and tissue remodeling. Mutations in the gene can result in coronary heart disease 6.
Specifications

Specifications

MMP3
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P03957, P08254, P28862
Mmp3
MMP3 Fusion Protein Ag12359
20 μL
Primary
Rat, Pig, Mouse, Human
Antibody
IgG1
Western Blot, Immunohistochemistry (Paraffin)
1F5A9
Unconjugated
MMP3
CHDS6, Matrix metalloproteinase 3, MMP 3, MMP3, SL 1, STMY, STMY1, STR1, Stromelysin 1, Transin 1
Mouse
Protein G
RUO
171045, 17392, 396769, 4314
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.