missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP3 Monoclonal Antibody (SL-1 IID4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA514210
This item is not returnable.
View return policy
Description
MA5-14210 targets MMP-3 (Stromelysin-1) in IF and WB applications and shows reactivity with Human samples. The MA5-14210 immunogen is aPMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1).
MMP3 (stromelysin-1) belongs to the peptidase M10A subfamily. It is secreted in its pro-protein form, and is activated by extracellular proteinase cleavage. MMP3 is capable of binding to zinc and calcium ions, and has metallo-endopeptidase activity. The active enzyme degrades collagen, gelatin, fibronectin, and laminin. It is involved in normal cellular processes such as wound repair and tissue remodeling. Mutations in the gene can result in coronary heart disease 6.
Specifications
| MMP3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P08254, P28862 | |
| MMP3 | |
| APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1) | |
| 500 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Western Blot | |
| SL-1 IID4 | |
| Unconjugated | |
| MMP3 | |
| CHDS6; EC 3.4.24.17; EMS-2; matrix metallo protease; matrix metallopeptidase 3; matrix metallopeptidase 3 (stromelysin 1, progelatinase); matrix metalloproteinase 3; matrix metalloproteinase 3 (stromelysin 1, progelatinase); matrix metalloproteinase 3 precursor; matrix metalloproteinase-3; metalloproteinase 3 receptor; MMP; MMP10; MMP3; MMP-3; MMP-3 protein; MMPs; progelatinase; proteoglycanase; SL-1; SLN1; SLN-1; STMY; STMY1; Str1; STR-1; stromelysin; stromelysin 1; Stromelysin1; stromelysin-1; Transin1; Transin-1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 17392, 4314 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction