missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP26 Antibody (S01-7A4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19696-50ul
This item is not returnable.
View return policy
Description
MMP26 Monoclonal antibody specifically detects MMP26 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MMP26 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S01-7A4 | |
| Unconjugated | |
| EC 3.4.24.-, Endometase, matrilysin 2, matrilysin-2, matrix metallopeptidase 26, matrix metalloproteinase 26, matrix metalloproteinase-26, MGC126590, MGC126592, MMP-26 | |
| A synthetic peptide of human MMP26 (Uniprot # Q3MJ78) | |
| 50 μg | |
| Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 56547 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction