missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MMP25 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5109967

Product Code. 17197893

  • £438.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SPRPARLHRF WEGLPAQVRV VQAAYARHRD GRILLFSGPQ FWVFQDRQLE GGARPLTELG LPPGEEVDAV FSWPQNGKTY LVRGRQYWRY DEAAARPDPG YPRDLSLWEG APPSPDDVTV SNAGDTYFFK GAHYWRFPKN SIKTEPDAPQ PMGPNWLDCP APSSGPRAPR PPKATPVSET CDCQCELNQA A.

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMPs are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. However, the protein encoded by this gene is a member of the membrane-type MMP (MT-MMP) subfamily, attached to the plasma membrane via a glycosylphosphatidyl inositol anchor. In response to bacterial infection or inflammation, the encoded protein is thought to inactivate alpha-1 proteinase inhibitor, a major tissue protectant against proteolytic enzymes released by activated neutrophils, facilitating the transendothelial migration of neutrophils to inflammatory sites. The encoded protein may also play a role in tumor invasion and metastasis through activation of MMP2. The gene has previously been referred to as MMP20 but has been renamed MMP25.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MMP25
Polyclonal
Unconjugated
Mmp25
F730048C11Rik; Leukolysin; matrix metallo protease; matrix metallopeptidase 25; matrix metallopeptidase-like 1; matrix metalloproteinase 20; matrix metalloproteinase 25; matrix metalloproteinase-25; matrix metalloproteinase-like 1; membrane-type 6 matrix metalloproteinase; membrane-type matrix metalloproteinase 6; Membrane-type-6 matrix metalloproteinase; MMP; MMP 25; MMP20; MMP20A; Mmp25; MMP-25; MMPL1; MMPs; MT MMP 6; MT6 MMP; MT6MMP; MT6-MMP; MT-MMP 6; MTMMP6; MT-MMP6
Rabbit
Affinity chromatography
RUO
240047, 302963, 64386
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot, Immunocytochemistry
1.05 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q3U435, Q9NPA2
Mmp25
Recombinant fusion protein containing a sequence corresponding to amino acids 350-540 of human MMP25 (NP_0719131).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.