missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP23B Antibody (1D8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00008510-M02
This item is not returnable.
View return policy
Description
MMP23B Monoclonal antibody specifically detects MMP23B in Human samples. It is validated for ELISA, ELISA
Specifications
| MMP23B | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| EC 3.4.24.-, Femalysin, matrix metallopeptidase 23B, matrix metalloproteinase 22, matrix metalloproteinase 23B, matrix metalloproteinase in the female reproductive tract, Matrix metalloproteinase-21, Matrix metalloproteinase-22, matrix metalloproteinase-23, MIFR, MIFR-1MMP22femalysin, MMP21, MMP-21, MMP-22, MMP-23, MMP23A | |
| MMP23B (NP_008914, 241 a.a. ~ 340 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LSQDELWGLHRLYGCLDRLFVCASWARRGFCDARRRLMKRLCPSSCDFCYEFPFPTVATTPPPPRTKTRLVPEGRNVTFRCGQKILHKKGKVYWYKDQEP | |
| 0.1 mg | |
| Signal Transduction | |
| 8510 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| ELISA, Sandwich ELISA | |
| 1D8 | |
| ELISA, Sandwich ELISA | |
| NP_008914 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction