missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MMP23 (Cleaved Tyr79) Polyclonal Antibody
GREENER_CHOICE

Product Code. 15554391
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15554391 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15554391 Supplier Invitrogen™ Supplier No. PA537434

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 37 and 44 kDa. Purity is >95% by SDS-PAGE.

Matrix metalloproteinases (MMPs) are highly homologous Zn2+ endopeptidases involved in extracellular matrix breakdown. MMP mediated extracellular remodeling occurs in normal physiological processes, such as embryonic development, reproduction and tissue remodeling, and disease processes, including arthritis and metastasis. MMP-23 exhibits sequence similarity with most MMPs, but displays a difference in domain structure. The MMP-23 protein contains prepro-, catalytic, cysteine-rich, Interleukin-1 receptor-related and proline-rich domains. Lacking a recognizable signal sequence, MMP-23 has a short prodomain. In addition, MMP-23 contains a single cysteine residue that can be part of the cysteine-switch mechanism operation for maintaining enzyme latency. MMP-23 is a membrane-anchored glycoprotein with type II topology. Subcellular localization is predominantly perinuclear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP23 (Cleaved Tyr79)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MMP23B
Gene Accession No. O75900
Gene Alias Femalysin; matrix metallopeptidase 23A (pseudogene); matrix metallopeptidase 23B; matrix metalloproteinase 22; matrix metalloproteinase 23B; matrix metalloproteinase in the female reproductive tract; matrix metalloproteinase-21; Matrix metalloproteinase-22; matrix metalloproteinase-23; Matrix metalloproteinase-23, soluble form; MIFR; MIFR-1; MMP 23; MMP21; MMP-21; MMP22; MMP-22; MMP23; MMP-23; MMP23A; MMP23B; soluble form
Gene Symbols MMP23B
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 51-100 of human MMP-23.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8510, 8511
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.