missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55467-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to MMP20.
MMP20 degrades amelogenin, the major protein component of the enamel matrix, as well as two of the macromolecules characterizing the cartilage extracellular matrix: aggrecan and the cartilage oligomeric matrix protein (COMP). MMP20 may play a central role in tooth enamel formation.Specifications
| MMP20 | |
| Polyclonal | |
| Unconjugated | |
| MMP20 | |
| Enamel metalloproteinase, Enamelysin, matrix metallopeptidase 20, Matrix metalloproteinase 20, MMP 20, MMP20 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 300341, 30800, 9313 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.74 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60882, P57748 | |
| MMP20 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction