missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2B10D1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66366-1-IG-150UL
This item is not returnable.
View return policy
Description
MMP (matrix metalloproteinase) are proteolytic enzymes capable of degrading connective tissue components. MMP have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPa and TIMPs play a significant role in regulating angiogenesis. MMP2 is synthesized as a 631 amino acid proenzyme which is activated by cleavage of the first 80 amino acids, and contains the basic structure of propeptide, catalytic, and hemopexin domains. The matrix metalloproteinases (MMPs) are a family of at least eighteen secreted and membrane-bound zincendopeptidases. Collectively, these enzymes can degrade all the components of the extracellular matrix, including fibrillar and non-fibrillar collagens, fibronectin, laminin and basement membrane glycoproteins. In general, a signal peptide, a propeptide, and a catalytic domain containing the highly conserved zinc-binding site characterizes the structure of the MMPs. Functionally, MMP2 is involved in tissue remodeling. Mutations in MMP-2 gene have been associated with Winchester syndrome and Nodulosis-Arthropathy-Osteolysis (NAO) syndrome. Two transcript variants encoding different isoforms of MMP-2 have been found.Specifications
| MMP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08253, P33434, P33436 | |
| Mmp2 | |
| MMP2 Fusion Protein Ag25039 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2B10D1 | |
| Unconjugated | |
| MMP2 | |
| 72 kDa gelatinase, 72 kDa type IV collagenase, CLG4, CLG4A, Gelatinase A, Matrix metalloproteinase 2, MMP 2, MMP II, MMP2, MONA, TBE 1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 17390, 397391, 4313, 81686 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction