Learn More
MMP19 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA124424
Description
PA1-24424 detects MMP-19 in human samples. PA1-24424 has been successfully used in Western blot and immunohistochemistry procedures. The PA1-24424 immunogen is a synthetic peptide region of human MMP-19. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling. They are also involved in disease processes, such as arthritis and metastasis. Most MMP′s are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The function of the protein encoded by this gene has not been determined. This gene was previously referred to as MMP18 but has been renamed matrix metalloproteinase 19 (MMP19). Multiple transcript variants encoding distict isoforms have been identified for this gene.
Specifications
| MMP19 | |
| Polyclonal | |
| Unconjugated | |
| Q99542 | |
| MMP19 | |
| Synthetic peptide corresponding to a region of human MMP-19. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| MMP19 | |
| MMP18, RASI-1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4327 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.