missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MMP11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579678
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat spleen tissue, MM231 whole cell. IHC: mouse spleen tissue, rat spleen tissue, human appendicitis tissue.
Stromelysin, a member of the matrix metalloproteinase family, demonstrates wide substrate specificity with the ability to degrade proteoglycan, fibronectin, laminin, casein, and the nonhelical region of collagen. The stromelysin-3 (MMP-11) gene was originally identified on the basis that it is expressed specifically in stromal cells surrounding invasive breast carcinomas. MMP-11 is localized by in situ hybridization to the long arm of chromosome 22. The MMP-11 expression is found more specifically in malignant tumors than in benign ones.
Specifications
| MMP11 | |
| Polyclonal | |
| Unconjugated | |
| Mmp11 | |
| matrix metallo protease; matrix metallopeptidase 11; matrix metallopeptidase 11 (stromelysin 3); Matrix metalloproteinase 11 (stromelysin 3); matrix metalloproteinase-11; MMP; MMP11; MMP-11; MMPs; SL-3; ST3; Stmy3; stromelysin III; stromelysin-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17385, 25481, 4320 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P24347, Q02853, Q499S5 | |
| Mmp11 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human MMP11 (104-135aa RWEKTDLTYRILRFPWQLVQEQVRQTMAEALK). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction