missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MMP11 Monoclonal Antibody (135421)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561334

Product Code. 30534228

  • £382.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Stromelysin, a member of the matrix metalloproteinase family, demonstrates wide substrate specificity with the ability to degrade proteoglycan, fibronectin, laminin, casein, and the nonhelical region of collagen. The stromelysin-3 (MMP-11) gene was originally identified on the basis that it is expressed specifically in stromal cells surrounding invasive breast carcinomas. MMP-11 is localized by in situ hybridization to the long arm of chromosome 22. The MMP-11 expression is found more specifically in malignant tumors than in benign ones.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MMP11
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P24347
Mmp11
E. coli-derived recombinant human MMP-11, Met1-Leu488.
100 μg
Primary
Human
Antibody
IgG2b
Flow Cytometry
135421
Unconjugated
Mmp11
matrix metallo protease; matrix metallopeptidase 11; matrix metallopeptidase 11 (stromelysin 3); Matrix metalloproteinase 11 (stromelysin 3); matrix metalloproteinase-11; MMP; MMP11; MMP-11; MMPs; SL-3; ST3; Stmy3; stromelysin III; stromelysin-3
Mouse
Protein A/G
RUO
4320
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.