missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP10 Monoclonal Antibody (IVC5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA514233
This item is not returnable.
View return policy
Description
MA5-14233 targets MMP-10 (Stromelysin-2) in IP and WB applications and shows reactivity with Human samples. The MA5-14233 immunogen is human recombinant MMP-10 (stromelysin-2 protein).
MMP10 belongs to the matrix metalloproteinase family, and is generally secreted in its pro form, which is further modified by extracellular proteinases. As a metalloproteinase, MMP10 requires zinc and calcium as cofactors for activity. It is involved in the activation of pro-collagenase, catabolism of collagen, degradation of fibronectin, and tissue remodeling. In humans, the gene MMP10 located on chromosome 11.
Specifications
| MMP10 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P09238 | |
| MMP10 | |
| Human recombinant MMP-10 (stromelysin-2 protein) | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| IVC5 | |
| Unconjugated | |
| MMP10 | |
| AV377895; matrix metallo protease; matrix metallopeptidase 10; matrix metallopeptidase 10 (stromelysin 2); matrix metalloprotease 10; matrix metalloproteinase 10; matrix metalloproteinase 10 (stromelysin 2); matrix metalloproteinase-10; MMP; Mmp10; MMP-10; MMPs; SL-2; STMY2; stromelysin 2; stromelysin-2; Transformation-associated protein 34A; transin 2; transin-2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 4319 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction