missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MMP10 Monoclonal Antibody (117239)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561991
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
MMP10 belongs to the matrix metalloproteinase family, and is generally secreted in its pro form, which is further modified by extracellular proteinases. As a metalloproteinase, MMP10 requires zinc and calcium as cofactors for activity. It is involved in the activation of pro-collagenase, catabolism of collagen, degradation of fibronectin, and tissue remodeling. In humans, the gene MMP10 located on chromosome 11.
Specifications
| MMP10 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P09238 | |
| MMP10 | |
| Mouse myeloma cell line NS0-derived recombinant human MMP-10, Tyr18-Cys476. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization, Western Blot | |
| 117239 | |
| Unconjugated | |
| MMP10 | |
| AV377895; matrix metallo protease; matrix metallopeptidase 10; matrix metallopeptidase 10 (stromelysin 2); matrix metalloprotease 10; matrix metalloproteinase 10; matrix metalloproteinase 10 (stromelysin 2); matrix metalloproteinase-10; MMP; Mmp10; MMP-10; MMPs; SL-2; STMY2; stromelysin 2; stromelysin-2; Transformation-associated protein 34A; transin 2; transin-2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 4319 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction