missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MMP1 Monoclonal Antibody (K29028_6B4), Invitrogen™

Product Code. 30195295
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30195295 200 μL 200µg
30207993 50 μL 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30195295 Supplier Invitrogen™ Supplier No. MA556182

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence of this protein is as follows: QTHLTYRIEN YTPDLPRADV DHAIEKAFQL WSNVTPLTFT KVSEGQADIM ISFVRGDHRD NSPFDGPGGN LAHAFQPGPG IGGDAHFDED ERWTNNFREY NLHRVAAHEL GHSLGLSHST DIGALMYPSY TFSGDVQLAQ DDIDGIQAIY GRSQNPVQPI GPQTPKACDS KLTFDAITTI

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. This gene encodes a secreted enzyme which breaks down the interstitial collagens, types I, II, and III. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K29028_6B4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene MMP1
Gene Accession No. P03956
Gene Alias 22 kDa interstitial collagenase; 27 kDa interstitial collagenase; CLG; CLGN; Collagenase; collagenase-like A; fibroblast collagenase; Interstitial collagenase; interstitial collagenase A; matrix metallo protease; matrix metallopeptidase 1; matrix metallopeptidase 1 (interstitial collagenase); matrix metallopeptidase 1a; matrix metallopeptidase 1a (interstitial collagenase); matrix metalloprotease 1; matrix metalloproteinase 1; matrix metalloproteinase 1a (interstitial collagenase); Matrix metalloproteinase1; Matrix metalloproteinase-1; Matrix metalloproteinase-1a; McolA; Mcol-A; MMP; MMP1; MMP-1; Mmp1a; MMP-1a; MMPs
Gene Symbols MMP1
Host Species Mouse
Immunogen Recombinant human MMP1 (immunogen range is 111-290).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4312
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.