missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MMP-9 Antibody (Yeda patent anti-MMP-9) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28864-1mg
This item is not returnable.
View return policy
Description
MMP-9 Monoclonal antibody specifically detects MMP-9 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| MMP-9 | |
| Monoclonal | |
| Unconjugated | |
| P14780 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| Yeda patent anti-MMP-9 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase | |
| MMP9 | |
| 1 mg | |
| Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR | |
| 4318 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction