missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-9 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
£191.00
Specifications
| Antigen | MMP-9 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1.25 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:250 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
MMP-9 Polyclonal specifically detects MMP-9 in Human, Mouse, Rat, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MMP-9 | |
| Western Blot 1.25 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:250 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| P14780 | |
| 4318 | |
| Synthetic peptide corresponding to the N-terminal of the human MMP-9 protein (between amino acids 150-200) UniProt P14780. | |
| Primary | |
| Reconstitute in 100 μL distilled water to a final concentration of 1mg/mL in PBS buffer. | |
| Expected identity based on immunogen sequence: Xenopus: 85%. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR | |
| PBS with 0.02% Sodium Azide | |
| 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase | |
| MMP9 | |
| IgG | |
| Immunogen affinity purified | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| 78 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title