missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-9 Antibody (4A3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 16 publications
£183.00 - £397.00
Specifications
| Antigen | MMP-9 |
|---|---|
| Clone | 4A3 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18030086
|
Novus Biologicals
NBP2-13173SS |
0.025 mL |
£183.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18751034
|
Novus Biologicals
NBP2-13173 |
£397.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
MMP-9 Monoclonal specifically detects MMP-9 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MMP-9 | |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR | |
| P14780 | |
| 4318 | |
| Synthetic peptide from the N-terminus of human MMP9 [Swiss-Prot# P14780] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 4A3 | |
| Western Blot 1:500 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase | |
| MMP9 | |
| IgG1 κ | |
| Protein G purified | |
| Active MMP9. It does not react with the MMP9 proenzyme or the active and proenzyme forms of MMP2. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title