missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-8 Antibody (MM0023-7A11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60989CL1
This item is not returnable.
View return policy
Description
MMP-8 Monoclonal antibody specifically detects MMP-8 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| MMP-8 | |
| Monoclonal | |
| CoraFluor 1 | |
| CLG1HNC, EC 3.4.24, EC 3.4.24.34, matrix metallopeptidase 8 (neutrophil collagenase), matrix metalloproteinase 8 (neutrophil collagenase), Matrix metalloproteinase-8, MMP-8, neutrophil collagenase, PMNL collagenase, PMNL-CL | |
| Human recombinant MMP-8 | |
| 0.1 mL | |
| Cancer | |
| 4317 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| MM0023-7A11 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction