missing translation for 'onlineSavingsMsg'
Learn More

MMP-8 Antibody (400102) [DyLight 755], Novus Biologicals™

Product Code. 30738098 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30738098 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738098 Supplier Novus Biologicals Supplier No. FAB3245Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MMP-8 Monoclonal antibody specifically detects MMP-8 in Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-8
Applications Western Blot
Classification Monoclonal
Clone 400102
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias CLG1HNC, EC 3.4.24, EC 3.4.24.34, matrix metallopeptidase 8 (neutrophil collagenase), matrix metalloproteinase 8 (neutrophil collagenase), Matrix metalloproteinase-8, MMP-8, neutrophil collagenase, PMNL collagenase, PMNL-CL
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant rat MMP-8, Leu21-Pro466, Accession # O88766
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4317
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.