missing translation for 'onlineSavingsMsg'
Learn More

MMP-8 Antibody (206), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18406777 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406777 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406777 Supplier Novus Biologicals Supplier No. NBP289361MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MMP-8 Monoclonal antibody specifically detects MMP-8 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-8
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 206
Conjugate mFluor Violet 500 SE
Dilution ELISA, Sandwich ELISA Capture
Formulation 50mM Sodium Borate
Gene Alias CLG1HNC, EC 3.4.24, EC 3.4.24.34, matrix metallopeptidase 8 (neutrophil collagenase), matrix metalloproteinase 8 (neutrophil collagenase), Matrix metalloproteinase-8, MMP-8, neutrophil collagenase, PMNL collagenase, PMNL-CL
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human MMP-8 (Accession#: NP_002415.1; Met1-Gly467).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4317
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.