missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-7 Antibody (704202) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB7170JF635
This item is not returnable.
View return policy
Description
MMP-7 Monoclonal antibody specifically detects MMP-7 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| MMP-7 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| EC 3.4.24, EC 3.4.24.23, matrilysin, matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase | |
| Mouse myeloma cell line NS0-derived recombinant mouse MMP-7, Met1-Leu264, Accession # Q10738 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 704202 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Wnt Signaling Pathway | |
| 4316 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction