missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-3 Rabbit anti-Human, Clone: MMP3/1994R, Novus Biologicals™
Rabbit Monoclonal Antibody
£412.00
Specifications
| Antigen | MMP-3 |
|---|---|
| Clone | MMP3/1994R |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, ELISA |
| Applications | Western Blot, Flow Cytometry, ELISA |
Description
MMP-3 Monoclonal specifically detects MMP-3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| MMP-3 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Extracellular Matrix | |
| 10 mM PBS with 0.05% BSA | |
| 4314 | |
| IgG | |
| Protein A purified |
| MMP3/1994R | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, stromelysin-1, transin-1 | |
| Recombinant fragment (around aa 317-327) of human MMP-3 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title