missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-3, Mouse anti-Human, Clone: MMP3/2655, Novus Biologicals™
Mouse Monoclonal Antibody
£211.00 - £377.00
Specifications
| Antigen | MMP-3 |
|---|---|
| Clone | MMP3/2655 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 ug/ml, ELISA, Protein Array |
| Applications | Western Blot, ELISA, Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18200118
|
Novus Biologicals
NBP2-79730-20UG |
20 μg |
£211.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18233118
|
Novus Biologicals
NBP2-79730-100UG |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MMP-3 Monoclonal specifically detects MMP-3 in Human samples. It is validated for Western Blot, ELISA, Protein Array.Specifications
| MMP-3 | |
| 0.2 mg/mL | |
| Western Blot, ELISA, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Extracellular Matrix | |
| 10 mM PBS with 0.05% BSA | |
| CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, stromelysin-1, transin-1 | |
| MMP3 | |
| IgG2b κ | |
| Protein A purified |
| MMP3/2655 | |
| Western Blot 1-2 ug/ml, ELISA, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P08254, P08254 | |
| 4314 | |
| Recombinant fragment of human MMP-3 protein (around aa 236-363) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title