missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-3 Antibody (MMP3/2806), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08626AF700
This item is not returnable.
View return policy
Description
MMP-3 Monoclonal antibody specifically detects MMP-3 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein Array
Specifications
| MMP-3 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, stromelysin-1, transin-1 | |
| Recombinant full-length human MMP-3 protein (Uniprot: P08254 ) | |
| 100 μg | |
| Angiogenesis, Cancer, Extracellular Matrix | |
| 4314 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| ELISA, Immunohistochemistry (Paraffin), Protein Array | |
| MMP3/2806 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido