missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-3 Antibody (50647), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB513C
This item is not returnable.
View return policy
Description
MMP-3 Monoclonal antibody specifically detects MMP-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| MMP-3 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 50647 | |
| Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, stromelysin-1, transin-1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human MMP-3, Tyr18-Cys477, Accession # P08254 | |
| 0.5 mL | |
| Angiogenesis, Cancer, Extracellular Matrix | |
| 4314 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction