missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse MMP-24/MT5-MMP Antibody, R&D Systems™

Description
MMP-24/MT5-MMP Monoclonal specifically detects MMP-24/MT5-MMP in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
Specifications
| Antigen | MMP-24/MT5-MMP |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | 143908 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | Q9R0S2 |
| Gene Alias | EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 24 (membrane-inserted), matrix metalloproteinase 24 (membrane-inserted), matrix metalloproteinase-24, membrane-type 5 matrix metalloproteinase, Membrane-type matrix metalloproteinase 5, Membrane-type-5 matrix metalloproteinase, MMP24, MMP-24, MT5MMP, MT5-MMP, MT5-MMPMMP25, MT-MMP 5, MTMMP5, MT-MMP5 |
| Gene Symbols | MMP24 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?