missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-24/MT5-MMP Antibody (RM0289-7J71), Alexa Fluor™ 750, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-11803AF750
This item is not returnable.
View return policy
Description
MMP-24/MT5-MMP Monoclonal specifically detects MMP-24/MT5-MMP in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| MMP-24/MT5-MMP | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MMP24 | |
| Mouse recombinant MMP-24/MT5-MMP (extracellular domain) | |
| 0.1 mL | |
| 10893.0 | |
| Mouse | |
| IgG2 |
| Western Blot, Flow Cytometry | |
| RM0289-7J71 | |
| Western Blot, Flow Cytometry | |
| EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 24 (membrane-inserted), matrix metalloproteinase 24 (membrane-inserted), matrix metalloproteinase-24, membrane-type 5 matrix metalloproteinase, Membrane-type matrix metalloproteinase 5, Membrane-type-5 matrix metalloproteinase, MMP-24, MT5MMP, MT5-MMPMMP25, MT-MMP 5, MTMMP5, MT-MMP5 | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| This antibody weakly cross reacts with mouse MMP3 and MMP-9. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction