missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-2 Antibody (8B4), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB200-114R
This item is not returnable.
View return policy
Description
MMP-2 Monoclonal specifically detects MMP-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.
Specifications
| MMP-2 | |
| Monoclonal | |
| DyLight 550 | |
| P08253 | |
| MMP2 | |
| Activated recombinant human MMP-2. [Uniprot: P08253] | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Cancer, Cell Biology, Cytoskeleton Markers, Extracellular Matrix, HIF Target Genes, Hypoxia, Tumor Suppressors | |
| 4313 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), KnockDown | |
| 8B4 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, KnockDown Validated | |
| 72 kDa gelatinase, CLG4, CLG4A72 kDa type IV collagenase, collagenase type IV-A, EC 3.4.24, EC 3.4.24.24, Gelatinase A, matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase), matrix metalloproteinase 2 (gelatinase A, 72kD gelatinase, 72kD type IVcollagenase), Matrix metalloproteinase-2, matrix metalloproteinase-II, MMP-2, MMP-II, MONA, neutrophil gelatinase, TBE-1matrix metalloproteinase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase) | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This is specific for pro and active MMP2. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction