missing translation for 'onlineSavingsMsg'
Learn More

MMP-2 Antibody (666047) [DyLight 650], Novus Biologicals™

Product Code. 30736273 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736273 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736273 Supplier Novus Biologicals Supplier No. FAB9024W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MMP-2 Monoclonal antibody specifically detects MMP-2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-2
Applications ELISA
Classification Monoclonal
Clone 666047
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias 72 kDa gelatinase, CLG4, CLG4A72 kDa type IV collagenase, collagenase type IV-A, EC 3.4.24, EC 3.4.24.24, Gelatinase A, matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase), matrix metalloproteinase 2 (gelatinase A, 72kD gelatinase, 72kD type IVcollagenase), Matrix metalloproteinase-2, matrix metalloproteinase-II, MMP-2, MMP-II, MONA, neutrophil gelatinase, TBE-1matrix metalloproteinase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase)
Host Species Mouse
Immunogen Chinese hamster ovary cell line, CHO-derived recombinant human MMP-2, Ile34-Cys660, Accession # P08253
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, HIF Target Genes, Hypoxia, Metastasis, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4313
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.