missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-2 Antibody (1A10), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB903I
This item is not returnable.
View return policy
Description
MMP-2 Monoclonal antibody specifically detects MMP-2 in Human samples. It is validated for Western Blot, Flow Cytometry
Specifications
| MMP-2 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 72 kDa gelatinase, CLG4, CLG4A72 kDa type IV collagenase, collagenase type IV-A, EC 3.4.24, EC 3.4.24.24, Gelatinase A, matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase), matrix metalloproteinase 2 (gelatinase A, 72kD gelatinase, 72kD type IVcollagenase), Matrix metalloproteinase-2, matrix metalloproteinase-II, MMP-2, MMP-II, MONA, neutrophil gelatinase, TBE-1matrix metalloproteinase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase) | |
| Chinese hamster ovary cell line CHO-derived recombinant human MMP-2 | |
| 0.2 mL | |
| Angiogenesis, Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, HIF Target Genes, Hypoxia, Metastasis, Tumor Suppressors | |
| 4313 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry | |
| 1A10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction