missing translation for 'onlineSavingsMsg'
Learn More

MMP-2 (72kDa Collagenase IV) Ab-4 Mouse Monoclonal Antibody, Epredia™

Product Code. 12642357 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
Unit Size:
100µL
1mL
200µL
500µL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Conjugate Concentration unitSize
12642357 1 mL Unconjugated 200μg/mL 1mL
12622357 100 μL Biotin 200μg/mL 100µL
12632357 500 μL Biotin 200μg/mL 500µL
12612357 1 mL Biotin 200μg/mL 1mL
12652357 100 μL Unconjugated 200μg/mL 100µL
12672357 100 μL Unconjugated 1mg/mL 100µL
12682357 200 μL Unconjugated 1mg/mL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 12642357 Supplier Epredia Supplier No. MS806P

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Epredia™ MMP-2 (72kDa Collagenase IV) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

MMPs are proteolytic enzymes capable of degrading connective tissue components. They have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPs and TIMPs play a significant role in regulating angiogenesis. MMP-2; also known as 72kDa collagenase IV or gelatinase A is synthesized as a 631 amino acid proenzyme which is activated by cleavage of the first 80 amino acids.

Host Species: Mouse

Clone: A-Gel VC2

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human APMA-activated native 72kDa MMP-2

Molecular Weight: 72kDa (proform) and ∽66kDa (active form)

Positive Control: Placenta; Bladder, breast and ovarian CAs

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (not qualified)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-2 (72kDa Collagenase IV) Ab-4
Classification Monoclonal
Clone A-Gel VC2
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human APMA-activated native 72kDa MMP-2
Quantity 1 mL
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.