missing translation for 'onlineSavingsMsg'
Learn More

MMP-19 Antibody [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30732813 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732813 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732813 Supplier Novus Biologicals Supplier No. AF6790J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

MMP-19 Polyclonal antibody specifically detects MMP-19 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-19
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.24.-, matrix metallopeptidase 19, matrix metalloproteinase 18, matrix metalloproteinase 19, Matrix metalloproteinase RASI, Matrix metalloproteinase-18, matrix metalloproteinase-19, MMP-18, MMP18MMP-19, RASI, RASI-1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human MMP-19, Leu229-Tyr508, Accession # Q99542
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 4327
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.