missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-16/MT3-MMP Antibody (782005), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1785X
This item is not returnable.
View return policy
Description
MMP-16/MT3-MMP Monoclonal antibody specifically detects MMP-16/MT3-MMP in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| MMP-16/MT3-MMP | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4325 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 782005 | |
| Flow Cytometry, CyTOF-ready | |
| chromosome 8 open reading frame 57, DKFZp761D112, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 16 (membrane-inserted), matrix metalloproteinase 16 (membrane-inserted), matrix metalloproteinase-16, Membrane-type matrix metalloproteinase 3, Membrane-type-3 matrix metalloproteinase, MMP-16, MMPX2, MMP-X2, MT3MMP, MT3-MMPC8orf57, MT-MMP 3, MT-MMP2, MTMMP3, MT-MMP3, Putative transmembrane protein C8orf57 | |
| Chinese hamster ovary cell line CHO-derived recombinant human MMP-16/MT3-MMP, Ala32-Pro535, Accession # P51512 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction