missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-15/MT2-MMP Antibody (MM0028-5D11), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60994JF646
This item is not returnable.
View return policy
Description
MMP-15/MT2-MMP Monoclonal specifically detects MMP-15/MT2-MMP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MMP-15/MT2-MMP | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MMP15 | |
| Human recombinant MMP-15/MT2-MMP | |
| 0.1 mL | |
| Primary | |
| This antibody recognizes human MMP-15 in western blot without cross reactivity with human MMP-14/16. | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0028-5D11 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 15 (membrane-inserted), matrix metalloproteinase 15 (membrane-inserted), Membrane-type matrix metalloproteinase 2, Membrane-type-2 matrix metalloproteinase, MMP-15, MT2-MMPMT2MMP, MTMMP2MT-MMP 2, SMCP-2matrix metalloproteinase-15 | |
| Mouse | |
| Protein G purified | |
| Extracellular Matrix | |
| 4324 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction