missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-15/MT2-MMP Antibody (130522) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9161JF585
This item is not returnable.
View return policy
Description
MMP-15/MT2-MMP Monoclonal antibody specifically detects MMP-15/MT2-MMP in Human samples. It is validated for Immunohistochemistry,Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| MMP-15/MT2-MMP | |
| Monoclonal | |
| Janelia Fluor 585 | |
| EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 15 (membrane-inserted), matrix metalloproteinase 15 (membrane-inserted), Membrane-type matrix metalloproteinase 2, Membrane-type-2 matrix metalloproteinase, MMP-15, MT2-MMPMT2MMP, MTMMP2MT-MMP 2, SMCP-2matrix metalloproteinase-15 | |
| Mouse myeloma cell line NS0-derived recombinant human MMP-15, aa 132-625 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 130522 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.5 mL | |
| Extracellular Matrix | |
| 4324 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction