missing translation for 'onlineSavingsMsg'
Learn More

MMP-14/MT1-MMP Antibody (S05-6H5), Novus Biologicals™

Rabbit Monoclonal Antibody

£328.00 - £500.00

Specifications

Antigen MMP-14/MT1-MMP
Clone S05-6H5
Concentration 0.3 mg/mL
Dilution Western Blot, ELISA, Immunohistochemistry 1:50, Immunocytochemistry/ Immunofluorescence 1:10-1:2000, Immunoprecipitation, Immunohistochemistry-Paraffin, RNA Inhibition, Sandwich ELISA 1:100-1:2000
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18692854
View Documents
Novus Biologicals
NBP3-19491-100ul
100 μg
£500.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18696884
View Documents
Novus Biologicals
NBP3-19491-50ul
50 μg
£328.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MMP-14/MT1-MMP Monoclonal antibody specifically detects MMP-14/MT1-MMP in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), RNA Inhibition, Sandwich ELISA
Specifications

Specifications

MMP-14/MT1-MMP
0.3 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Rabbit
Cancer, Extracellular Matrix
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
4323
IgG
Affinity purified
S05-6H5
Western Blot, ELISA, Immunohistochemistry 1:50, Immunocytochemistry/ Immunofluorescence 1:10-1:2000, Immunoprecipitation, Immunohistochemistry-Paraffin, RNA Inhibition, Sandwich ELISA 1:100-1:2000
Monoclonal
Purified
RUO
Human, Mouse, Rat
EC 3.4.24, EC 3.4.24.80, matrix metallopeptidase 14 (membrane-inserted), matrix metalloproteinase 14 (membrane-inserted), matrix metalloproteinase-14, membrane type 1 metalloprotease, Membrane-type matrix metalloproteinase 1, Membrane-type-1 matrix metalloproteinase, MMP-14, MMP-X1, MT1MMP, MT1-MMPMTMMP1, MT-MMP 1
A synthetic peptide of human MMP-14/MT1-MMP (Uniprot # P50281)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.