missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-14/MT1-MMP Antibody (MM0027-9E10), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60993AF488
This item is not returnable.
View return policy
Description
MMP-14/MT1-MMP Monoclonal specifically detects MMP-14/MT1-MMP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MMP-14/MT1-MMP | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MMP14 | |
| Human recombinant MMP-14/MT1-MMP | |
| 0.1 mL | |
| Primary | |
| This antibody recognizes human MMP-14 in western blot without cross reactivity with human MMP-1-13, and MMP14-15. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0027-9E10 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 3.4.24, EC 3.4.24.80, matrix metallopeptidase 14 (membrane-inserted), matrix metalloproteinase 14 (membrane-inserted), matrix metalloproteinase-14, membrane type 1 metalloprotease, Membrane-type matrix metalloproteinase 1, Membrane-type-1 matrix metalloproteinase, MMP-14, MMP-X1, MT1MMP, MT1-MMPMTMMP1, MT-MMP 1 | |
| Mouse | |
| Protein G purified | |
| Cancer, Extracellular Matrix | |
| 4323 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction