missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-17310
This item is not returnable.
View return policy
Description
MMP-13 Polyclonal specifically detects MMP-13 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| MMP-13 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| CLG 3, CLG3EC 3.4.24, collagenase 3, EC 3.4.24.-, EC 3.4.24.22, EC 3.4.24.24, EC 3.4.24.35, EC 3.4.24.65, EC 3.4.24.7, MANDP1, matrix metallopeptidase 13 (collagenase 3), matrix metalloproteinase 13 (collagenase 3), Matrix metalloproteinase-13, MMP-13 | |
| Rabbit | |
| 54 kDa | |
| 0.1 mL | |
| Cancer, Cellular Markers, Extracellular Matrix | |
| 4322 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen Assay dependent | |
| P45452 | |
| MMP13 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human MMP13. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Rabbit (100%), Mouse (93%), Rat (93%), Sheep (93%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction