missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-11 Antibody (135421), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3657CL1
This item is not returnable.
View return policy
Description
MMP-11 Monoclonal antibody specifically detects MMP-11 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| MMP-11 | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 3.4.24, EC 3.4.24.-, EC 3.4.24.24, matrix metallopeptidase 11 (stromelysin 3), matrix metalloproteinase 11 (stromelysin 3), Matrix metalloproteinase-11, MMP-11, SL-3, ST3, STMY3stromelysin 3, stromelysin III, stromelysin-3 | |
| E. coli-derived recombinant human MMP-11, Met1-Leu488, Accession # P24347 | |
| 0.1 mL | |
| Angiogenesis, Cancer, Extracellular Matrix | |
| 4320 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 135421 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction