missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-10 Antibody (110316) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9102JF525
This item is not returnable.
View return policy
Description
MMP-10 Monoclonal antibody specifically detects MMP-10 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| MMP-10 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| EC 3.4.24, EC 3.4.24.22, matrix metallopeptidase 10 (stromelysin 2), matrix metalloprotease 10, matrix metalloproteinase 10 (stromelysin 2), Matrix metalloproteinase-10, MMP-10, SL-2, STMY2stromelysin-2, stromelysin 2, transin 2, transin-2 | |
| Mouse myeloma cell line NS0-derived recombinant human MMP-10, Phe99-Cys476, Accession # P09238 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 110316 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.5 mL | |
| Angiogenesis, Cancer, Extracellular Matrix | |
| 4319 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction