Learn More
MMP-1 Antibody (6A5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
£198.00 - £481.00
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|
18719503
|
Novus Biologicals
NBP2-22123SS |
£198.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18768793
|
Novus Biologicals
NBP2-22123 |
£481.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
MMP-1 Monoclonal specifically detects MMP-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.For Research Use Only