missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mMESSAGE mMACHINE™ T7 mRNA Kit with CleanCap™ Reagent AG

Product Code. 31662617
Change view
Click to view available options
No. of Reactions:
1000 Reactions
25 Reactions
50 Reactions
Unit Size:
1000 reactions
50 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
31662617 25 Reactions -
17904741 50 Reactions 50 reactions
17994731 1000 Reactions 1000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31662617 Supplier Invitrogen™ Supplier No. A5762025
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The mMESSAGE mMACHINE T7 mRNA Kit with CleanCap Reagent AG contains all the reagents needed to generate mRNA with naturally occurring Cap 1 structure while providing increased yields and higher quality with greater translatability in vivo compared to legacy capping methods such as ARCA caps.

The mMESSAGE mMACHINE T7 mRNA Kit with CleanCap Reagent AG contains all the reagents and buffers necessary for high quality in vitro synthesis of large amounts of mRNA with the naturally occurring Cap 1 structure. Utilization of the CleanCap Reagent AG ensures superior results relative to older capping technologies and greater protein production. Each standard 20-μL reaction yields a minimum of 100 μg of Cap 1-containing mRNA, while the scaled-up protocol yields up to 1 mg of mRNA. This kit is suitable for full- or partially-modified-nucleotide substitution to generate modified mRNA.

Features of the mMESSAGE mMACHINE T7 mRNA Kit with CleanCap Reagent AG include:

  • Contains proven CleanCap capping technology
  • Optimized to achieve high mRNA yields (>5 mg/mL)
  • Produces mRNA that is over 95% capped
  • Optimized for production of mRNA from 0.9–10 kb templates
  • Minimum yields of 100 μg per reaction, up to 150 μg per reaction (see figure below)
  • Allows use of modified nucleotides, full- or partial-modified substitution
  • Includes linearized 0.9 kb GFP control template, yields ≥100 μg mRNA
  • Made up of high quality TheraPure reagents to enable transition from RUO to GMP
  • Available in two sizes: 50 and 1000 reactions*

*Note: 1000 reaction kit is designed to support scale-up to large amounts of mRNA production enabling ≥100 mg mRNA.

The mMESSAGE mMACHINE T7 mRNA Kit with CleanCap Reagent AG contains the trinucleotide cap analog CleanCap Reagent AG, which has the Cap 1 structure and can be used for co-transcriptional capping of mRNA without compromising yield. This reagent produces high capping efficiencies compared to legacy capping reagents such as ARCA. Cap 1-containing mRNAs are considerably more stable and have superior translational activities compared to Cap 0 mRNAs produced using ARCA or mCap cap analogs.

A linearized DNA template containing an AG-initiating T7 RNA polymerase promoter is strongly recommended for this kit to achieve mRNA yields >5 mg/mL with over 90% of the transcripts capped. The mRNA produced with this kit can be used for many applications, including transfections, preclinical mRNA therapeutic mRNA studies, preclinical vaccine development, microinjections, in vitro translation, and RNA structure and function analysis.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type HEK293, 293, HeLa, CHO-K1, CHO, Jurkat, Primary Human T Cells, Primary Cells, Stem Cells, iPSC, NIH-3T3, HepG2, A549, HUVEC
Final Product Type Synthetic mRNA, mRNA
Format Animal Origin Free Kit
Isolation Technology Silica Spin Column
Purification Target mRNA, Synthetic mRNA
Sample Type mRNA, Synthetic mRNA
No. of Reactions 25 Reactions
Product Line mMESSAGE mMACHINE
Product Type T7 In Vitro Transcription kit
Quantity 25 Reactions
Promoter Modified T7
Shipping Condition Dry Ice
Yield >100 μg
Starting Material PCR Products, Linearized DNA Plasmid
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.