missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MMEL1 Monoclonal Antibody (2D2H5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531799
This item is not returnable.
View return policy
Description
MA5-31799 has been tested in indirect ELISA.
MMEL1 is a member of the neutral endopeptidase (NEP) or membrane metallo-endopeptidase (MME) family. Family members play important roles in pain perception, arterial pressure regulation, phosphate metabolism and homeostasis. MMEL1 is a type II transmembrane protein and is thought to be expressed as a secreted protein. The protein encoded by this gene is a member of the neutral endopeptidase (NEP) or membrane metallo-endopeptidase (MME) family. Family members play important roles in pain perception, arterial pressure regulation, phosphate metabolism and homeostasis. This protein is a type II transmembrane protein and is thought to be expressed as a secreted protein. This gene is expressed mainly in testis with weak expression in the brain, kidney, and heart.
Specifications
| MMEL1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q495T6 | |
| MMEL1 | |
| Purified recombinant fragment of human MMEL1 (AA: 1-107) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 2D2H5 | |
| Unconjugated | |
| MMEL1 | |
| EC 3.4.24; EC 3.4.24.11; mel transforming oncogene-like 1; Mell1; membrane metalloendopeptidase like 1; Membrane metallo-endopeptidase-like 1; Membrane metallo-endopeptidase-like 1, soluble form; Membrane metallo-endopeptidase-like 2; MGC119454; MGC119455; MGC119456; Mmel1; MMEL2; NEP2; NEP2(m); NEP2(s); NEPII; NEPIIMGC119456; NEPLP; NEPLP alpha; NEPLP beta; NEPLP gamma; neprilysin 2; neprilysin II; Neprilysin-2; Neprilysin-2 secreted; Neprilysin-like 1; neprilysin-like peptidase; Nl1; NL-1; NL2; NL2NL1; SEP; Soluble secreted endopeptidase; zinc metallopeptidase | |
| Mouse | |
| Protein G | |
| RUO | |
| 79258 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction