Learn More
MMD2 Polyclonal Antibody
GREENER_CHOICE
MMD2 Polyclonal Antibody
Brand:  Invitrogen PA598604
Product Code. 16591422
  • £425.00 / 100µg
Description
MMD2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MMD2
Polyclonal
Unconjugated
MMD2
4930518M15Rik; C88001; Mmd2; monocyte to macrophage differentiation associated 2; monocyte to macrophage differentiation factor 2; monocyte to macrophage differentiation-associated 2; monocyte-to-macrophage differentiation factor 2; monocyte-to-macrophage differentiation-associated protein 2; PAQR10; Progestin and adipoQ receptor family member 10; progestin and adipoQ receptor family member X
Rabbit
Protein G
RUO
100505381, 221938, 75104
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q8IY49, Q8R189
MMD2
Recombinant Human Monocyte to macrophage differentiation factor 2 protein (1-38AA).
100 μg
Primary
Human, Mouse