missing translation for 'onlineSavingsMsg'
Learn More

MMAF hydrochloride, MedChemExpress

Product Code. 16439207
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16439207

Brand: Medchem Express HY15579A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MMAF (Monomethylauristatin F) hydrochloride is a potent tubulin polymerization inhibitor and is used as a antitumor agent. MMAF hydrochloride is widely used as a cytotoxic component of antibody-drug conjugates (ADCs) such as Vorsetuzumab mafodotin and SGN-CD19A.

MMAF (Monomethylauristatin F) hydrochloride is a potent tubulin polymerization inhibitor and is used as a antitumor agent. MMAF hydrochloride is widely used as a cytotoxic component of antibody-drug conjugates (ADCs) such as Vorsetuzumab mafodotin and SGN-CD19A.

Specifications

Chemical Name or Material MMAF hydrochloride
Physical Form Solid
CAS 1415246-68-2
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C39H66ClN5O8
For Use With (Application) Cancer-programmed cell death
Synonym Monomethylauristatin F hydrochloride
Solubility Information DMSO : 27 mg/mL (35.14 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES CC(C)[C@H](NC)C(N[C@H](C(N([C@@H]([C@@H](C)CC)[C@H](OC)CC(N1CCC[C@@]1([H])[C@H](OC)[C@@H](C)C(N[C@H](C(O)=O)CC2=CC=CC=C2)=O)=O)C)=O)C(C)C)=O.Cl
Molecular Weight (g/mol) 768.42
Formula Weight 768.42
Percent Purity 97.71%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.