missing translation for 'onlineSavingsMsg'
Learn More

MMAE-d8, MedChemExpress

Product Code. p-200100115
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16491479

Brand: Medchem Express HY15162A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MMAE-d8 (Monomethyl auristatin E-d8) is a deuterated labeled MMAE, a potent mitotic inhibitor and a tubulin inhibitor.

MMAE-d8 (Monomethyl auristatin E-d8) is a deuterated labeled MMAE, a potent mitotic inhibitor and a tubulin inhibitor.

Specifications

Chemical Name or Material MMAE-d8
CAS 2070009-72-0
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C39H59D8N5O7
Technique Cancer-programmed Cell Death
Color White
Synonym Monomethyl auristatin E-d8 Deuterated labeled MMAE
Solubility Information DMSO : 100 mg/mL (137.74 mM; Need ultrasonic)
SMILES CC[C@H](C)[C@@H]([C@@H](CC(N1[C@@H](CCC1)[C@@H]([C@@H](C)C(N[C@H](C)[C@@H](O)C2=CC=CC=C2)=O)OC)=O)OC)N(C([C@@](C(C([2H])([2H])[2H])(C([2H])([2H])[2H])[2H])([2H])NC([C@H](C(C)C)NC)=O)=O)C
Molecular Weight (g/mol) 726.03
Quantity 1 mg
Formula Weight 726.03
Percent Purity 98.43%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.