missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MMADHC Monoclonal Antibody (OTI1G4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526465
This item is not returnable.
View return policy
Description
MMADHC Monoclonal Antibody for Western Blot
Involved in cobalamin metabolism.
Specifications
| MMADHC | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H3L0 | |
| MMADHC | |
| Human recombinant protein fragment corresponding to amino acids 39-296 of MMADHC produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1G4 | |
| Unconjugated | |
| MMADHC | |
| 2010311D03Rik; AI314967; C2orf25; CblD; CL25022; HSPC161; likely ortholog of H. sapiens chromosome 2 open reading frame 25 (C2orf25); methylmalonic aciduria (cobalamin deficiency) cblD type, with homocystinuria; methylmalonic aciduria and homocystinuria type D homolog, mitochondrial; Methylmalonic aciduria and homocystinuria type D protein, mitochondrial; methylmalonic aciduria and homocystinuria, cblD type; Mmadhc; My011; protein C2orf25, mitochondrial | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 27249 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction