missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Squalene, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Squalene is an intermediate product in the synthesis of cholesterol, and shows several pharmacological properties such as hypolipidemic, hepatoprotective, cardioprotective, antioxidant, and antitoxicant activity. Squalene also has anti-fungal activity and can be used for the research of Trichophyton mentagrophytes treatment.
Specifications
Specifications
| Chemical Name or Material | Squalene |
| Physical Form | Liquid |
| CAS | 111-02-4 |
| Color | Colorless |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C30H50 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | Super Squalene trans-Squalene AddaVax |
| Solubility Information | DMSO : 16.67 mg/mL (40.59 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction