missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MMACHC Polyclonal Antibody

Description
Immunogen sequence: LKPFLQSCHL RMLTDPVDQC VAYHLGRVRE SLPELQIEII ADYEVHPNRR PKILAQTAAH VAGAAYYYQR QDVEADPWGN QRISGVCIHP RFGGWFAIRG VVLLPGIEVP DLPPRKPHDC VPTRADRIAL LEGFNFHWRD WTYRDAVTPQ ERYSEEQKAY FSTPPAQRLA LLGLAQPSEK PSSPSPDLPF TTPAPKKPGN PSRARSWLSP RVSPPASPGP; Positive Samples: Raji, Jurkat, Mouse kidney, Rat brain; Cellular Location: Cytoplasm.
The exact function of the MMACHC protein is not known, however, its C-terminal region shows similarity to TonB, a bacterial protein involved in energy transduction for cobalamin (vitamin B12) uptake. Hence, it is postulated that this protein may have a role in the binding and intracellular trafficking of cobalamin. Mutations in this gene are associated with methylmalonic aciduria and homocystinuria type cblC.
Specifications
Specifications
| Antigen | MMACHC |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 3.89 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MMACHC |
| Gene Accession No. | Q9CZD0, Q9Y4U1 |
| Gene Alias | 1810037K07Rik; cblC; Cyanocobalamin reductase (cyanide-eliminating); methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria; methylmalonic aciduria and homocystinuria type C protein; Methylmalonic aciduria and homocystinuria type C protein homolog; methylmalonic aciduria cblC type, with homocystinuria; Mmachc; RGD1310806; RP11-291L19.3 |
| Gene Symbols | MMACHC |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?